2-Chloro-4-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

B23542-14 B23542-22
CAAAAB23542-14EA 56.56 CAD
CAAAAB23542-14 CAAAAB23542-22 CAAAAB23542-36
2-Chloro-4-nitrotoluene 98%
2-Chloro-4-nitrotoluene

Purity: 98% CAS No.: 121-86-8 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 62-66° Boiling Point: 260° UN No.: 3457 MDL No.: MFCD00007210

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 260 °C
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00007210
CAS Number: 121-86-8
EINECS: 204-501-7
UN: 3457
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR