(±)-Pipecolinic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23541-06
B23541-14
B23541-22
CAAAAB23541-06EA
62.57
CAD
CAAAAB23541-06
CAAAAB23541-14
CAAAAB23541-22
(±)-Pipecolinic acid 99%
(±)-Pipecolinic acid
Purity: 99%
CAS No.: 535-75-1
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 280° dec.
Beilstein Reference: 81095
Merck Reference: 14,7458
MDL No.: MFCD00064347
Formula:
C₆H₁₁NO₂ Melting Pt: ∼ 280 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064347 CAS Number: 535-75-1 EINECS: 208-616-3 Merck Index: 14,7458 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...