2,3,5-Collidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23537-06
B23537-14
B23537-22
CAAAAB23537-06EA
63.04
CAD
CAAAAB23537-06
CAAAAB23537-14
CAAAAB23537-22
2,3,5-Collidine 99%
2,3,5-Collidine
Purity: 99%
CAS No.: 695-98-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 74°(165°F)
Refractive Index: 1.5075
Odor: Unpleasant
Boiling Point: 182-184°
Beilstein Reference: 108832
UN No.: 2810
MDL No.: MFCD00126840
Formula:
C₈H₁₁N Boiling Pt: 182…184 °C Density: 0.935 Flash Pt: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00126840 CAS Number: 695-98-7 EINECS: 211-786-1 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...