2-Ethylhexyl bromide 96% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

B23532-14 B23532-22
CAAAAB23532-14EA 42.98 CAD
CAAAAB23532-14 CAAAAB23532-22 CAAAAB23532-36
2-Ethylhexyl bromide 96% stabilized
2-Ethylhexyl bromide

Purity: 96% CAS No.: 18908-66-2 Molecular Formula: C8H17Br Formula Weight: 193.13 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Refractive Index: 1.4538 Boiling Point: 94-95°/35mm Beilstein Reference: 1098313 MDL No.: MFCD00000220

Formula: C₈H₁₇Br
MW: 193.13 g/mol
Boiling Pt: 94…95 °C (35 mmHg)
Density: 1.086
Flash Pt: 69 °C (156 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000220
CAS Number: 18908-66-2
EINECS: 242-659-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR