tert-Butyl-4-methoxyphenol (mixture of isomers) 96%

Supplier: Thermo Scientific Chemicals
Warning

B23530-0B B23530-18 B23530-30
CAAAAB23530-0BEA 508.76 CAD
CAAAAB23530-0B CAAAAB23530-18 CAAAAB23530-30
tert-Butyl-4-methoxyphenol (mixture of isomers) 96%
tert-Butyl-4-methoxyphenol (mixture of isomers)

Antioxidant


Purity: 96% CAS No.: 25013-16-5 Molecular Formula: C11H16O2 Formula Weight: 180.25 Melting Point: 55-65° Merck Reference: 14,1547 MDL No.: MFCD01779059

Formula: C₁₁H₁₆O₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 55…65 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD01779059
CAS Number: 25013-16-5
EINECS: 246-563-8
Merck Index: 14,1547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR