2-Fluoro-5-(trifluoromethyl)anisole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23516-03
B23516-06
CAAAAB23516-03EA
37.6
CAD
CAAAAB23516-03
CAAAAB23516-06
2-Fluoro-5-(trifluoromethyl)anisole 97%
2-Fluoro-5-(trifluoromethyl)anisole
Purity: 97%
CAS No.: 261951-78-4
Molecular Formula: C8H6F4O
Formula Weight: 194.13
Flash Point: 62°(144°F)
Refractive Index: 1.4352
MDL No.: MFCD01631564
Formula:
C₈H₆F₄O MW: 194.13 g/mol Boiling Pt: 151 Melting Pt: -15 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01631564 CAS Number: 261951-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...