3-Bromophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride

B23510-06 B23510-14 B23510-22
CAAAAB23510-06EA 156.1 CAD
CAAAAB23510-06 CAAAAB23510-14 CAAAAB23510-22
3-Bromophenylhydrazinium chloride 98%
3-Bromophenylhydrazinium chloride

Purity: 98% CAS No.: 27246-81-7 Molecular Formula: C6H7BrN2.HCl Formula Weight: 223.51 Melting Point: ca 225° dec. Beilstein Reference: 3565829

Formula: C₆H₈BrClN₂
MW: 223.5 g/mol
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012933
CAS Number: 27246-81-7
EINECS: 248-362-0
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR