Troclosene sodium, (max. 3% H₂O) 97% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

B23504-14 B23504-22 B23504-36
CAAAAB23504-14EA 47.87 CAD
CAAAAB23504-14 CAAAB23504-22 CAAAB23504-36
Troclosene sodium, (max. 3% H₂O) 97% (dry weight)
Troclosene sodium

Purity: 97% CAS No.: 2893-78-9 Molecular Formula: C3Cl2N3NaO3 Formula Weight: 219.95 Odor: Chlorine-like Melting Point: ca 250° dec. Beilstein Reference: 4056155 UN No.: 2465 MDL No.: MFCD00006036

Formula: C₃Cl₂N₃NaO₃
Melting Pt: ca 250 °C (dec.)
Density: 0.96 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006036
CAS Number: 2893-78-9
EINECS: 220-767-7
UN: 2465
ADR: "5.1",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR