1-Chloro-2,5-dimethoxybenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23497-18
B23497-30
CAAAAB23497-18EA
60.67
CAD
CAAAAB23497-18
CAAAAB23497-30
1-Chloro-2,5-dimethoxybenzene 99%
1-Chloro-2,5-dimethoxybenzene
Purity: 99%
CAS No.: 2100-42-7
Molecular Formula: C8H9ClO2
Formula Weight: 172.61
Flash Point:
Formula:
C₈H₉ClO₂ Boiling Pt: 231…234 °C Melting Pt: 8,1 °C Density: 1,210 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008356 CAS Number: 2100-42-7 EINECS: 218-267-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...