ω-Chloro-3,4-dihydroxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23496-14
B23496-22
B23496-36
CAAAAB23496-14EA
87.85
CAD
CAAAAB23496-14
CAAAAB23496-22
CAAAAB23496-36
ω-Chloro-3,4-dihydroxyacetophenone 97%
ω-Chloro-3,4-dihydroxyacetophenone
Purity: 97%
CAS No.: 99-40-1
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
Melting Point: 172-176°
Beilstein Reference: 2092660
Formula:
C₈H₇ClO₃ Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002200 CAS Number: 99-40-1 EINECS: 202-754-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...