2,3,5,6-Tetrafluorothiophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Mercapto-2,3,5,6-tetrafluorobenzene, 2,3,5,6-Tetrafluorobenzenethiol, 2,3,5,6-Tetrafluorobenzene-1-thiol, 2,3,5,6-tetrafluorothiophenol

B23483-06 B23483-14
CAAAB23483-06EA 172.22 CAD
CAAAB23483-06 CAAAB23483-14
2,3,5,6-Tetrafluorothiophenol 98%
2,3,5,6-Tetrafluorothiophenol

Purity: 98% CAS No.: 769-40-4 Molecular Formula: C6H2F4S Formula Weight: 182.14 Flash Point: 48°(119°F) Refractive Index: 1.4870 Odor: Stench Boiling Point: 152-153° UN No.: 1992 MDL No.: MFCD00004829

Formula: C₆H₂F₄S
MW: 182.14 g/mol
Boiling Pt: 152…153 °C
Density: 1.53
Flash Pt: 48 °C (119 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004829
CAS Number: 769-40-4
EINECS: 212-210-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR