Acenaphthenequinone 95%

Supplier: Thermo Scientific Chemicals
Warning

B23466-06 B23466-14 B23466-22
CAAAAB23466-06EA 54.83 CAD
CAAAAB23466-06 CAAAAB23466-14 CAAAAB23466-22
Acenaphthenequinone 95%
Acenaphthenequinone

Purity: 95% CAS No.: 82-86-0 Molecular Formula: C12H6O2 Formula Weight: 182.18 Melting Point: 253-262° Beilstein Reference: 879172

Formula: C₁₂H₆O₂
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 253…262 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003805
CAS Number: 82-86-0
EINECS: 201-441-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR