4-Amino-3-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyano-2-nitroaniline

B23463-03 B23463-06 B23463-14
CAAAAB23463-03EA 77.74 CAD
CAAAAB23463-03 CAAAAB23463-06 CAAAAB23463-14
4-Amino-3-nitrobenzonitrile 98%
4-Amino-3-nitrobenzonitrile

Purity: 98% CAS No.: 6393-40-4 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 159-161° UN No.: 3439 MDL No.: MFCD00013373

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00013373
CAS Number: 6393-40-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR