Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23459-09
B23459-18
CAAAAB23459-09EA
91.64
CAD
CAAAAB23459-09
CAAAAB23459-18
Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)
Patton-Reeders reagent (Calconcarboxylic acid)
CAS No.: 3737-95-9
Molecular Formula: C21H14N2O7S
Formula Weight: 438.42
Melting Point: ca 300° dec.
Beilstein Reference: 727492
MDL No.: MFCD00004078
Formula:
C₂₁H₁₄N₂O₇S Melting Pt: ∼ 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004078 CAS Number: 3737-95-9 EINECS: 223-117-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...