α,α,α-Trifluoro-o-xylene 97%

Supplier: Thermo Scientific Chemicals
Warning

B23453-03 B23453-06
CAAAB23453-03EA 84.21 CAD
CAAAB23453-03 CAAAB23453-06
α,α,α-Trifluoro-o-xylene 97%
α,α,α-Trifluoro-o-xylene

Purity: 97% CAS No.: 13630-19-8 Molecular Formula: C8H7F3 Formula Weight: 160.14 Refractive Index: 1.4350 Boiling Point: 125-126° UN No.: 1993 MDL No.: MFCD00153204

Formula: C₈H₇F₃
Boiling Pt: 125…126 °C
Density: 1.174
Storage Temperature: Ambient
MDL Number: MFCD00153204
CAS Number: 13630-19-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR