Aminomethanesulfonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23441-22
B23441-36
CAAAB23441-22EA
142.2
CAD
CAAAB23441-22
CAAAB23441-36
Aminomethanesulfonic acid 97%
Aminomethanesulphonic acid
Purity: 97%
CAS No.: 13881-91-9
Molecular Formula: H2NCH2SO3H
Formula Weight: 111.12
pH: 3.2
Melting Point: ca 180° dec.
Beilstein Reference: 1811756
UN No.: 2585
MDL No.: MFCD00008124
Formula:
CH₅NO₃S Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008124 CAS Number: 13881-91-9 EINECS: 237-649-6 UN: 2585 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...