3,5-Dimethylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

B23434-03 B23434-06 B23434-14
CAAAAB23434-03EA 69.68 CAD
CAAAAB23434-03 CAAAAB23434-06 CAAAAB23434-14
3,5-Dimethylphenylboronic acid 98%
3,5-Dimethylphenylboronic acid

Purity: 98% CAS No.: 172975-69-8 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: 235-237° Beilstein Reference: 7368658

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR