3-Fluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

B23426-06 B23426-14 B23426-22
CAAAB23426-06EA 76.47 CAD
CAAAB23426-06 CAAAB23426-14 CAAAB23426-22
3-Fluorobenzonitrile 98%
3-Fluorobenzonitrile

Purity: 98% CAS No.: 403-54-3 Molecular Formula: C7H4FN Formula Weight: 121.11 Flash Point: 67°(152°F) Refractive Index: 1.5055 Odor: Almond-like Melting Point: -16° Boiling Point: 183° Beilstein Reference: 1858941 UN No.: 3276 MDL No.: MFCD00001797

Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 183 °C
Melting Pt: –16 °C
Density: 1.140
Flash Pt: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR