2-Cyclopenten-1-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone
B23408-06
B23408-14
CAAAB23408-06EA
118.82
CAD
CAAAB23408-06
CAAAB23408-14
2-Cyclopenten-1-one 98%
2-Cyclopenten-1-one
Purity: 98%
CAS No.: 930-30-3
Molecular Formula: C5H6O
Formula Weight: 82.10
Storage Temperature: 2°C to 8°C
Flash Point: 42°(108°F)
Refractive Index: 1.4810
Odor: Characteristic
Boiling Point: 150°
UN No.: 1224
MDL No.: MFCD00001401
Formula:
C₅H₆O MW: 82.1 g/mol Boiling Pt: 150 °C Density: 1.01 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001401 CAS Number: 930-30-3 EINECS: 213-213-0 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...