3-Methyl-3-buten-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23398-AC
B23398-AE
CAAAB23398-ACEA
39.66
CAD
CAAAB23398-AC
CAAAB23398-AE
3-Methyl-3-buten-1-ol 97%
3-Methyl-3-buten-1-ol
Purity: 97%
CAS No.: 763-32-6
Molecular Formula: CH2=C(CH3)CH2CH2OH
Formula Weight: 86.13
Flash Point: 36°(97°F)
Refractive Index: 1.4330
Boiling Point: 130-133°
Beilstein Reference: 1071239
UN No.: 1987
MDL No.: MFCD00002933
Formula:
C₅H₁₀O Boiling Pt: 130…133 °C Melting Pt: –82 °C Density: 0.850 Flash Pt: 36 °C (97 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002933 CAS Number: 763-32-6 EINECS: 212-110-8 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...