3-(Methylthio)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

B23184-14 B23184-22 B23184-06
CAAAAB23184-14EA 715.74 CAD
CAAAAB23184-14 CAAAAB23184-22 CAAAB23184-06
3-(Methylthio)aniline 97%
3-(Methylthio)aniline

Purity: 97% CAS No.: 1783-81-9 Molecular Formula: C7H9NS Formula Weight: 139.23 Flash Point: 110°(230°F) Refractive Index: 1.6370 Odor: Unpleasant Boiling Point: 163-165°/16mm Beilstein Reference: 2078599 UN No.: 2810 MDL No.: MFCD00007793

Formula: C₇H₉NS
Boiling Pt: 163…165 °C/16 mm
Density: 1.130
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007793
CAS Number: 1783-81-9
EINECS: 217-262-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR