Dipropyl sulfide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23155-14
B23155-22
B23155-36
CAAAB23155-14EA
125.61
CAD
CAAAB23155-14
CAAAB23155-22
CAAAB23155-36
Dipropyl sulfide 98+%
Dipropyl sulphide
Purity: 98+%
CAS No.: 111-47-7
Molecular Formula: (CH3CH2CH2)2S
Formula Weight: 118.24
Flash Point: 28°(82°F)
Refractive Index: 1.4487
Odor: Stench
Melting Point: -102°
Boiling Point: 141-142°
Beilstein Reference: 1719002
Merck Reference: 14,7868
UN No.: 1993
MDL No.: MFCD00009379
Formula:
C₆H₁₄S Boiling Pt: 141…142 °C Melting Pt: –102 °C Density: 0.838 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009379 CAS Number: 111-47-7 EINECS: 203-873-8 UN: 1993 ADR: 3,III Merck Index: 14,7868 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...