Terephthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

B23137-14 B23137-22
CAAAB23137-14EA 52.3 CAD
CAAAB23137-14 CAAAB23137-22
Terephthalonitrile 98%
Terephthalonitrile

Purity: 98% CAS No.: 623-26-7 Molecular Formula: C8H4N2 Formula Weight: 128.13 Odor: Odorless Melting Point: 224-227° Beilstein Reference: 1072210 MDL No.: MFCD00001810

Formula: C₈H₄N₂
MW: 128.13 g/mol
Melting Pt: 224…227 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001810
CAS Number: 623-26-7
EINECS: 210-783-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR