3-Fluorothiophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

B23114-03 B23114-06 B23114-14
CAAAB23114-03EA 66.04 CAD
CAAAB23114-03 CAAAB23114-06 CAAAB23114-14
3-Fluorothiophenol 98%
3-Fluorothiophenol

Purity: 98% CAS No.: 2557-77-9 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 58°(136°F) Refractive Index: 1.5540 Odor: Stench Boiling Point: 156° Beilstein Reference: 2039302 UN No.: 3071 MDL No.: MFCD00040227

Formula: C₆H₅FS
Boiling Pt: 156 °C
Density: 1.203
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040227
CAS Number: 2557-77-9
EINECS: 219-876-2
UN: 1228
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR