Methyltrichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro(methyl)silane

B23107-22 B23107-36
CAAAB23107-22EA 58.93 CAD
CAAAB23107-22 CAAAB23107-36
Methyltrichlorosilane 97%
Methyltrichlorosilane

Purity: 97% CAS No.: 75-79-6 Molecular Formula: CH3Cl3Si Formula Weight: 149.48 Flash Point: 8°(46°F) Refractive Index: 1.4115 Odor: Pungent Melting Point: -78° Boiling Point: 64-66° Beilstein Reference: 1361381 UN No.: 1250 MDL No.: MFCD00000481

Formula: CH₃Cl₃Si
MW: 149.48 g/mol
Boiling Pt: 64…66 °C
Melting Pt: –78 °C
Density: 1.270
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000481
CAS Number: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,II
REACH: 01-2119475226-36

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR