Creatinine 98%
Supplier: Thermo Scientific Chemicals
B23097-09
B23097-18
B23097-30
CAAAB23097-09EA
50.72
CAD
CAAAB23097-09
CAAAB23097-18
CAAAB23097-30
Creatinine 98%
Creatinine
Purity: 98%
CAS No.: 60-27-5
Molecular Formula: C4H7N3O
Formula Weight: 113.12
Odor: Odorless
pH: 7.5-8.5
Melting Point: ca 255° dec.
Beilstein Reference: 112061
Merck Reference: 14,2569
MDL No.: MFCD00059730
Formula:
C₄H₇N₃O Melting Pt: ca 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00059730 CAS Number: 60-27-5 EINECS: 200-466-7 Merck Index: 14,2569 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...