Isobutanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

B23091-0F B23091-AP
CAAAAB23091-0FEA 257.54 CAD
CAAAAB23091-0F CAAAAB23091-AP
Isobutanol 99%
Isobutanol

Purity: 99% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Refractive Index: 1.3960 Odor: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740

Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107…108 °C
Melting Pt: –108 °C
Density: 0.802
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 14,5131
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR