4-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

B23072-03 B23072-06
CAAAB23072-03EA 83.27 CAD
CAAAB23072-03 CAAAB23072-06
4-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%
4-Methoxy-3-(trifluoromethyl)benzoyl chloride

Purity: 97% CAS No.: 98187-18-9 Molecular Formula: C9H6ClF3O2 Formula Weight: 238.59 Refractive Index: 1.5150

Formula: C₉H₆ClF₃O₂
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD01631576
CAS Number: 98187-18-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR