3-Aminopropan-1-ol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23041-22
B23041-36
CAAAB23041-22EA
45.19
CAD
CAAAB23041-22
CAAAB23041-36
3-Aminopropan-1-ol 99%
3-Aminopropan-1-ol
Purity: 99%
CAS No.: 156-87-6
Molecular Formula: H2N(CH2)3OH
Formula Weight: 75.11
Flash Point: 101°(214°F)
Refractive Index: 1.4610
Odor: Amine-like
Melting Point: 11°
Boiling Point: 184-187°
Beilstein Reference: 741855
UN No.: 2735
MDL No.: MFCD00008223
Formula:
C₃H₉NO Boiling Pt: 184…187 °C Melting Pt: 11 °C Density: 0.983 Flash Pt: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008223 CAS Number: 156-87-6 EINECS: 205-864-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...