1,5-Diaminopentane 98%

Supplier: Thermo Scientific Chemicals
Danger

B23039-06 B23039-14
CAAAB23039-06EA 195.92 CAD
CAAAB23039-06 CAAAB23039-14
1,5-Diaminopentane 98%
1,5-Diaminopentane

Purity: 98% CAS No.: 462-94-2 Molecular Formula: C5H14N2 Formula Weight: 102.18 Refractive Index: 1.4585 Odor: Amine-like Boiling Point: 178-180° Merck Reference: 14,1609 UN No.: 2735 MDL No.: MFCD00008239

Formula: C₅H₁₄N₂
Boiling Pt: 178…180 °C
Melting Pt: 9 °C
Density: 0.873
Storage Temperature: Ambient
MDL Number: MFCD00008239
CAS Number: 462-94-2
EINECS: 207-329-0
UN: 2735
ADR: 8,III
Merck Index: 14,1609

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR