2,4-Dinitrobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B23035-06 B23035-14 B23035-22
CAAAB23035-06EA 97.33 CAD
CAAAB23035-06 CAAAB23035-14 CAAAB23035-22
2,4-Dinitrobenzenesulfonyl chloride 98%
2,4-Dinitrobenzenesulphonyl chloride

Purity: 98% CAS No.: 1656-44-6 Molecular Formula: C6H3ClN2O6S Formula Weight: 266.62 Melting Point: 100-104° UN No.: 3096 MDL No.: MFCD00007431

Formula: C₆H₃ClN₂O₆S
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00007431
CAS Number: 1656-44-6
EINECS: 216-749-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR