p-Toluonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23029-0B
B23029-18
B23029-30
CAAAB23029-0BEA
584.6
CAD
CAAAB23029-0B
CAAAB23029-18
CAAAB23029-30
p-Toluonitrile 98+%
p-Toluonitrile
Purity: 98+%
CAS No.: 104-85-8
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 85°(185°F)
Odor: Almond-like
Boiling Point: 217°
Beilstein Reference: 507386
Merck Reference: 14,9538
MDL No.: MFCD00001827
Formula:
C₈H₇N Boiling Pt: 217 °C Melting Pt: 26…29 °C Density: 0.981 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001827 CAS Number: 104-85-8 EINECS: 203-244-8 Merck Index: 14,9538 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...