Hexanoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23027-22
B23027-36
CAAAB23027-22EA
79.79
CAD
CAAAB23027-22
CAAAB23027-36
Hexanoyl chloride 98%
Hexanoyl chloride
Purity: 98%
CAS No.: 142-61-0
Molecular Formula: CH3(CH2)4COCl
Formula Weight: 134.61
Flash Point: 50°(122°F)
Refractive Index: 1.4260
Odor: Acrid
Melting Point: -87°
Boiling Point: 150-153°
Merck Reference: 14,1763
UN No.: 2920
MDL No.: MFCD00000760
Formula:
C₆H₁₁ClO Boiling Pt: 150…153 °C Melting Pt: –87 °C Density: 0.971 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000760 CAS Number: 142-61-0 EINECS: 205-549-1 UN: 2920 ADR: 8,II Merck Index: 14,1763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...