Sebaconitrile 95%

Supplier: Thermo Scientific Chemicals
Danger

B23010-06 B23010-14
CAAAB23010-06EA 43.13 CAD
CAAAB23010-06 CAAAB23010-14 CAAAB23010-22
Sebaconitrile 95%
Sebaconitrile

Purity: 95% CAS No.: 1871-96-1 Molecular Formula: NC(CH2)8CN Formula Weight: 164.25 Flash Point: 110°(230°F) Melting Point: 4-8° Boiling Point: 199-200° Beilstein Reference: 1750739 MDL No.: MFCD00019900

Formula: C₁₀H₁₆N₂
Boiling Pt: 199…200 °C
Melting Pt: 4…8 °C
Density: 0.910
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019900
CAS Number: 1871-96-1
EINECS: 217-494-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR