3,5-Dimethoxybenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

B22989-06 B22989-14
CAAAB22989-06EA 96.06 CAD
CAAAB22989-06 CAAAB22989-14 CAAAB22989-22
3,5-Dimethoxybenzonitrile 98%
3,5-Dimethoxybenzonitrile

Purity: 98% CAS No.: 19179-31-8 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 87-89° UN No.: 3439 MDL No.: MFCD00001804

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001804
CAS Number: 19179-31-8
EINECS: 242-858-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR