Pivalic anhydride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylacetic anhydride, 2,2-Dimethylpropionic anhydride

B22983-14 B22983-22 B22983-36
CAAAB22983-14EA 99.07 CAD
CAAAB22983-14 CAAAB22983-22 CAAAB22983-36
Pivalic anhydride 99%
Pivalic anhydride

Purity: 99% CAS No.: 1538-75-6 Molecular Formula: C10H18O3 Formula Weight: 186.25 Flash Point: 63°(145°F) Refractive Index: 1.4090 Odor: Unpleasant Boiling Point: 193° Beilstein Reference: 386552 UN No.: 3265 MDL No.: MFCD00008842

Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
Boiling Pt: 193 °C
Density: 0.918
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008842
CAS Number: 1538-75-6
EINECS: 216-263-1
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR