(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B22968-ME B22968-77
CAAAAB22968-MEEA 141.25 CAD
CAAAAB22968-ME CAAAB22968-77
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

Purity: 99% CAS No.: 20445-31-2 Molecular Formula: C10H9F3O3 Formula Weight: 234.18 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4730 Melting Point: 43-45° Boiling Point: 105-107°/1mm Beilstein Reference: 2215440 Merck Reference: 14,6280 Optical Rotation: +72° (c=2 in methanol) MDL No.: MFCD00004184

Formula: C₁₀H₉F₃O₃
Boiling Pt: 105…107 °C/1 mm
Melting Pt: 43…45 °C
Density: 1.344
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5
Merck Index: 14,6280

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR