trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate
Purity: 98%
CAS No.: 125572-95-4
Molecular Formula: C14H22N2O8.H2O
Formula Weight: 364.35 (346.34anhy)
Melting Point: ca 220° dec.
Beilstein Reference: 2226756
Formula:
C₁₄H₂₂N₂O₈·1H₂O MW: 364.35 g/mol Melting Pt: ∼ 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066429 CAS Number: 125572-95-4 EINECS: 236-308-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...