2,4-Lutidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-Dimethylpyridine
B22913-14
B22913-22
CAAAB22913-14EA
59.57
CAD
CAAAB22913-14
CAAAB22913-22
2,4-Lutidine 98+%
2,4-Lutidine
Purity: 98+%
CAS No.: 108-47-4
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 47°(117°F)
Refractive Index: 1.5000
Odor: Unpleasant
Melting Point: -60°
Boiling Point: 158-160°
UN No.: 1992
MDL No.: MFCD00006337
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 158…160 °C Melting Pt: –60 °C Density: 0.931 Flash Pt: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006337 CAS Number: 108-47-4 EINECS: 203-586-8 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...