α-Bromo-4-chlorotoulene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22858-06
B22858-14
CAAAB22858-06EA
81.69
CAD
CAAAB22858-06
CAAAB22858-14
CAAAB22858-22
α-Bromo-4-chlorotoulene 98+%
α-Bromo-4-chlorotoulene
Purity: 98+%
CAS No.: 622-95-7
Molecular Formula: C7H6BrCl
Formula Weight: 205.49
Odor: Pungent
Melting Point: 46-51°
Boiling Point: 107-108°/8mm
Beilstein Reference: 606497
UN No.: 3261
MDL No.: MFCD00040714
Formula:
C₇H₆BrCl Boiling Pt: 107…108 °C/8 mm Melting Pt: 46…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040714 CAS Number: 622-95-7 EINECS: 210-760-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...