o-Tolyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

B22845-06 B22845-14 B22845-22
CAAAB22845-06EA 74.1 CAD
CAAAB22845-06 CAAAB22845-14 CAAAB22845-22
o-Tolyl isothiocyanate 98%
o-Tolyl isothiocyanate

Purity: 98% CAS No.: 614-69-7 Molecular Formula: C8H7NS Formula Weight: 149.22 Flash Point: 107°(224°F) Refractive Index: 1.6361 Odor: Pungent Boiling Point: 239° Beilstein Reference: 878487 UN No.: 2922 MDL No.: MFCD00004802

Formula: C₈H₇NS
MW: 149.22 g/mol
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004802
CAS Number: 614-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR