4-Amino-2,2-difluoro-1,3-benzodioxole 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22797-03
B22797-06
CAAAB22797-03EA
225.94
CAD
CAAAB22797-03
CAAAB22797-06
4-Amino-2,2-difluoro-1,3-benzodioxole 97+%
4-Amino-2,2-difluoro-1,3-benzodioxole
Purity: 97+%
CAS No.: 106876-54-4
Molecular Formula: C7H5F2NO2
Formula Weight: 173.12
UN No.: 2810
MDL No.: MFCD00792416
Formula:
C₇H₅F₂NO₂ MW: 173.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792416 CAS Number: 106876-54-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...