4-(Trifluoromethyl)benzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22745-06
B22745-14
CAAAAB22745-06EA
153.58
CAD
CAAAAB22745-06
CAAAAB22745-14
4-(Trifluoromethyl)benzylamine 97%
4-(Trifluoromethyl)benzylamine
Purity: 98%
CAS No.: 3300-51-4
Molecular Formula: C8H8F3N
Formula Weight: 175.15
Flash Point: 75°(167°F)
Refractive Index: 1.4640
Boiling Point: 80-82°/15mm
Beilstein Reference: 2640698
UN No.: 2735
MDL No.: MFCD00010220
Formula:
C₈H₈F₃N MW: 175.15 g/mol Boiling Pt: 80…82 °C/15 mm Density: 1.229 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010220 CAS Number: 3300-51-4 EINECS: 221-971-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...