Piperidin-4-ol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22723-06
B22723-14
B22723-22
CAAAB22723-06EA
44.71
CAD
CAAAB22723-06
CAAAB22723-14
CAAAB22723-22
Piperidin-4-ol 97%
Piperidin-4-ol
Purity: 97%
CAS No.: 5382-16-1
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 107°(224°F)
Odor: Amine-like
pH: 11.5-11.7
Melting Point: 86-90°
Boiling Point: 108-114°/10mm
Beilstein Reference: 102738
UN No.: 3263
MDL No.: MFCD00005999
Formula:
C₅H₁₁NO Boiling Pt: 108…114 °C/10 mm Melting Pt: 86…90 °C Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005999 CAS Number: 5382-16-1 EINECS: 226-373-1 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...