2-Amino-5-chlorobenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22690-03
B22690-06
CAAAB22690-03EA
39.03
CAD
CAAAB22690-03
CAAAB22690-06
2-Amino-5-chlorobenzonitrile 97%
2-Amino-5-chlorobenzonitrile
Purity: 97%
CAS No.: 5922-60-1
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 94-97°
Boiling Point: 132-135°/0.5mm
UN No.: 3439
MDL No.: MFCD00017106
Formula:
C₇H₅ClN₂ Boiling Pt: 132…135 °C/0.5 mm Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017106 CAS Number: 5922-60-1 EINECS: 227-651-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...