1-Chloro-2-propanol ≥75% (remainder mainly 2-chloro-1-propanol), Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloropropan-2-ol, Propylene chlorohydrin, Chloropropanol
B22686-14
B22686-22
CAAAB22686-14EA
63.83
CAD
CAAAB22686-14
CAAAB22686-22
1-Chloro-2-propanol ≥75% (remainder mainly 2-chloro-1-propanol), Technical Grade
1-Chloro-2-propanol
Purity: 75%
CAS No.: 127-00-4
Molecular Formula: CH3CH(OH)CH2Cl
Formula Weight: 94.54
Flash Point: 51°(123°F)
Refractive Index: 1.4370
Boiling Point: 126-127°
Beilstein Reference: 773653
Merck Reference: 14,7850
UN No.: 2929
MDL No.: MFCD00004530
Formula:
C₃H₇ClO Boiling Pt: 126…127 °C Density: 1.111 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004530 CAS Number: 127-00-4 EINECS: 204-819-6 UN: 2611 ADR: 6.1,II Merck Index: 14,7850 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...