3-Chloro-2-methylpropene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22674-0B
B22674-18
B22674-30
CAAAB22674-0BEA
409.22
CAD
CAAAB22674-0B
CAAAB22674-18
CAAAB22674-30
3-Chloro-2-methylpropene 98%
3-Chloro-2-methylpropene
Purity: 98%
CAS No.: 563-47-3
Molecular Formula: C4H7Cl
Formula Weight: 90.55
Flash Point: -12°(9°F)
Refractive Index: 1.4275
Melting Point: -80°
Boiling Point: 71-72°
Merck Reference: 14,2148
UN No.: 2554
MDL No.: MFCD00000953
Formula:
C₄H₇Cl Boiling Pt: 71…72 °C Melting Pt: –80 °C Density: 0.917 Flash Pt: –12 °C (9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000953 CAS Number: 563-47-3 EINECS: 209-251-2 UN: 2554 ADR: 3,II Merck Index: 14,2148 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...