3-Chlorothiophene 98% may contain up to 2% DMF
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22669-06
B22669-14
B22669-22
CAAAB22669-06EA
74.58
CAD
CAAAB22669-06
CAAAB22669-14
CAAAB22669-22
3-Chlorothiophene 98% may contain up to 2% DMF
3-Chlorothiophene
Purity: 98%
CAS No.: 17249-80-8
Molecular Formula: C4H3ClS
Formula Weight: 118.59
Flash Point: 37°(98°F)
Refractive Index: 1.5520
Odor: Unpleasant
Boiling Point: 137-139°
Beilstein Reference: 105255
UN No.: 1992
MDL No.: MFCD00043887
Formula:
C₄H₃ClS MW: 118.59 g/mol Boiling Pt: 137…139 °C Density: 1.276 Flash Pt: 37 °C (98 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043887 CAS Number: 17249-80-8 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...