4-Methyl-2-pentenoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22639-09
B22639-18
CAAAB22639-09EA
83.9
CAD
CAAAB22639-09
CAAAB22639-18
4-Methyl-2-pentenoic acid 98+%
4-Methyl-2-pentenoic acid
Purity: 98+%
CAS No.: 10321-71-8
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point:
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 212 °C Density: 0.959 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043804 CAS Number: 10321-71-8 EINECS: 233-706-4 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...