4-Fluorophthalonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Fluorobenzene-1,2-dicarbonitrile, 4-Fluoro-1,2-benzenedicarbonitrile, 4-Fluoro-1,2-dicyanobenzene
B22624-03
B22624-06
CAAAAB22624-MDEA
102.92
CAD
CAAAAB22624-MD
CAAAB22624-03
CAAAB22624-06
4-Fluorophthalonitrile 98%
4-Fluorophthalonitrile
Purity: 98%
CAS No.: 65610-14-2
Molecular Formula: C8H3FN2
Formula Weight: 146.12
Melting Point: 100-102°
Beilstein Reference: 2443236
UN No.: 3439
MDL No.: MFCD00210430
Formula:
C₈H₃FN₂ MW: 146.12 g/mol Boiling Pt: 294 °C (1013 hPa) Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210430 CAS Number: 65610-14-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...